VWR® Film Seals for Fluorescence and Photoprotection, Black
# de catalogue 89087-692
Fournisseur: VWR International
Spécifications
- Optical property:Light blocking
- Sterility:Non sterile
- Description:Black vinyl films
- Thickness:240,0 µm
- Cat. no.:89087-692
Spécifications
A propos de cet article
These films are 240 µm thick black vinyl with adhesive for use with 96-well, 384-well, and 1536-well black microplates. They fit within the edges of raised-rim plates and may be applied to either the plate top or bottom.
- Edge cutouts assist in positioning around ribs on the plate bottom
- Split backing aids in applying the film properly
- No end tabs
- Recommended temperature range −40 to +80 °C (-40 to 176 °F)
The light-blocking and absorbing properties of these films make them ideal for reducing stray light and crosstalk in fluorescence applications and for protecting light-sensitive samples in storage.
Each film L×D: 114×76 mm
Customers Who Bought This Also Bought