VWR® PCR Films, PE
# de catalogue 60941-122
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Polyethylene Films
- L×W:13×7.6 cm (51/8×3")
- Thickness:76 µm
- Cat. no.:60941-122
Spécifications
A propos de cet article
These medium-density polyethylene films, 76µm thick with a 46µm thick adhesive layer, are designed for general-purpose microplate sealing and PCR.
- Pierceable
- Nonsterile
Films are pierceable and supplied nonsterile.
Customers Who Bought This Also Bought