VWR® Aluminium Foil Seals for Cold Storage
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without generation of aerosols or cross-contamination.
- Recommended for temperatures from −80 to +120 °C
- Not recommended for PCR
Each foil L×D: 133×82 mm (51/4×31/4")
Ordering information: Films are not recommended for PCR or thermocycling applications. Sterile product is packaged in bags of 50.
Customers Who Bought This Also Bought