VWR® Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Aluminium foils, 75.7 µm thick, for use with 96- or 384-well plates. Fit inside the rim of raised rim plates. These foils have one partial-width end tab. Available non sterile only.
- Recommended for temperatures from −80 to +120 °C
- Certified free from DNase, RNase and nucleic acids
Customers Who Bought This Also Bought