KIMBLE® HI-VAC® Low Holdup Valves, Straight, with PTFE Plug, without Tip O-Ring, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Sidearms are bent to minimize holdup that normally occurs in conventional valves
PTFE shaft seals protect the Viton® O-rings from contact with the system. Seal design reduces outgassing.
Customers Who Bought This Also Bought