KIMBLE® HI-VAC® Low Holdup Valves, Straight, with PTFE Plug and Extended Tip, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Sidearms are bent to minimize holdup of liquids
O-rings are made of Viton® rubber.
Customers Who Bought This Also Bought