8293 P100 Particulate Respirator, 3M™
# de catalogue CA56221-028
Fournisseur: 3M
Spécifications
- Description:8293 P100 Particulate Respirator
- Cat. no.:CA56221-028
- Supplier No.:70070757474
Spécifications
A propos de cet article
This respirator is well suited for those who want NIOSH's highest rated filtration efficiency in a maintenance-free respirator. For workplace applications involving OSHA substance-specific particle exposures including lead, cadmium, and arsenic.
- For workplace applications involving OSHA substance-specific particle exposures including lead, cadmium, and arsenic
- 3M Cool Flow® exhalation valve, a comfortable faceseal, and adjustable straps
- Provides minimum filtration efficiency of 99.97% against certain solid and liquid aerosols including oils
Respirator features the 3M Cool Flow® exhalation valve, a comfortable faceseal, and adjustable straps. Provides minimum filtration efficiency of 99.97% against certain solid and liquid aerosols including oils.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Customers Who Bought This Also Bought