OxiClear™ Disposable Inline Gas Purifiers
Fournisseur: Labclear
Certificats
A propos de cet article
Ideal for gas chromatography, glove boxes, and anaerobic experiments.
Oxygen-absorbing gas purifiers with high efficiency and low resistance to gas flow. Reduce amount of oxygen, trace amounts of water, and organic compounds from inert carrier gases such as nitrogen, helium, argon, and hydrogen. Tubular inline configuration with nickel-plated brass, threaded, male tube compression fitting on both ends.
Spécifications
- Catalog No:
- 32574-021
- 32574-054
- 32574-065
- 32574-076
- Max. operating pressure:
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- Maximum Flow Rate:
- 5 L/min. at 5 psi
- 5 L/min. at 5 psi
- 5 L/min. at 5 psi
- 5 L/min. at 5 psi
- Max. pressure:
- 250 psi
- 250 psi
- 250 psi
- 250 psi
- Oxygen removal:
- \<50 ppb
- \<50 ppb
- \<50 ppb
- \<50 ppb