InterMask™ Face Mask, InterWorld™
# de catalogue 46000-136
Fournisseur: Interworld
Spécifications
- Color:White
- Description:Face mask
- Size:Universal
- Cat. no.:46000-136
- Supplier No.:10070-115-02
Spécifications
A propos de cet article
Breathable mask is made of comfortable polypropylene materials and has three-ply construction for excellent filtration efficiency.
Ultrasonically welded polyester earloops provide a secure fit.
Ordering information: Packaged in a cleanroom environment 50 masks per bag, 20 bags per case.
Customers Who Bought This Also Bought