KIMTECH PURE® M3 Pleat-Style Face Masks, Kimberly-Clark Professional®
Fournisseur: Kimberly-Clark
CE Compliant
Certificats
A propos de cet article
Masks have an outer layer of apertured polyethylene film to provide protection from contamination.
- Outer layer of apertured polyethylene film
- Nonsterile face masks features nonirritating BICOSOF® inner facing and bendable aluminum nosepiece
- Sterile face masks features polyester binding and polyester filter media
Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments.
Nonsterile face masks offer excellent breathability with filtration of 99% at 0.1 µm. Sterile masks provides bacteria filtration efficiency (BFE) of 91.9% and particle filtration efficiency (PFE) of 93.1%. Sterile masks are gamma-sterilized to 10⁻⁶.
Ordering information: All masks are individually packaged. Packs are double bagged.
Customers Who Bought This Also Bought