Lint-Free Masks, Foamtec International WCC
Fournisseur: Foamtec International
Certificats
A propos de cet article
These fiber-free foam masks provide continuous particle/moisture control and cool, flexible comfort without the speech constraints, goggle/glasses steaming, or moisture build-up on the face typical of paper and dense cloth masks.
- Particle/moisture control restricts vapor transfer from breath to work surface
- Cool comfort even during strenuous production and maintenance procedures
- Easy to communicate and breathable making it ideal as an under mask for built-in fabric face veils and shields
- No restriction of breath or speech clarity; reduces lifting or pulling down masks
- Minimizes fogging of safety glasses/goggles
- 100% fiber-free
Foam masks are completely free of fibers and are non-shedding for use in cleanrooms where ultra tight particle
control is required. Ideal for applications where fibers from non-woven masks may not be tolerated.
Laundered and packaged in an ISO 5 / Class 100 Cleanroom.
Customers Who Bought This Also Bought