Cell Culture Aspiration Station, Welch®
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
Ideal for aqueous and bleach solutions, this station is an economical and portable solution for aspirating tissue culture samples
Note: Not for use with acidic, basic, or organic vapors and gases.
System is powered by a high-flow WOB-L piston pump that vacuums to 93mbar (70torr). Station accepts 7mm (1/4") I.D. tubing and automatically shuts down when the 1.2L (0.3gal.) collection receiver is full. Station features a hydrophobic inline filter, a vacuum regulator, and carrying handles. Bleach-resistant construction and corrosion-resistant wetted parts extend service life. Collection receiver is autoclavable.
Certifications: The 230V, 50Hz model is CE marked.
Ordering information: Supplied with a vacuum regulator, dial vacuum gauge, 1.2L (0.3gal.) autoclavable liquid trap, 1.8m (6') of tubing, and two hydrophobic filters.
Customers Who Bought This Also Bought