Compact Aspiration/Pressure Transfer Station, Welch®
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
Oil-free station is an economical and portable device for aspirating and filtering
Note: Not for use with acidic, basic, or organic vapors and gases.
System is ideal for aspirating aqueous solutions including buffers. Pump has a pressure capability to 33psig and vacuum capability to 292mbar (219torr). Station accepts 5mm (3/16") tubing and automatically shuts down when the 1.2L (0.3gal.) collection receiver is full. Station features a hydrophobic inline filter, a vacuum regulator, and carrying handles. All wetted parts are corrosion resistant. Collection receiver is autoclavable.
Certifications: The 230V, 50Hz model is CE marked.
Ordering information: Supplied with vacuum regulator, dial vacuum gauge, 1.2L (0.3gal.) autoclavable liquid trap, 1.8m (6') of tubing, and two hydrophobic filters.
Customers Who Bought This Also Bought