VACUSAFE Vacuum Aspiration System, INTEGRA Biosciences
Fournisseur: INTEGRA Biosciences
Certificats
A propos de cet article
The VACUSAFE™ Aspiration System combines vacuum pump, collection bottle, overflow protection, and filter into an all-in-one liquid aspiration system
This closed aspiration system offers the most effective combination of containment of hazardous liquids and maximum personal protection.
Asperation rate: 17 mL/s
Certifications: CE certified.
Ordering information: Complete aspiration system includes integrated vacuum pump, collection bottle, bottle handle, tubing, hydrophobic filter, mains adapter, and VACUBOY® hand operator (see 37001-682 series).
Spécifications
- Catalog No:
- 80076-628
- 89031-564
- 89031-562
- Dimensions:
- 530H×180W×320D mm (20.9×7.1×12.6")
- 530H×180W×320D mm (20.9×7.1×12.6")
- 530H×180W×320D mm (20.9×7.1×12.6")
- Input voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- Noise rating:
- <50 dB(A) at 1 m
- <50 dB(A) at 1 m
- <50 dB(A) at 1 m
- Pump flow rate:
- 8 L/min. (Air)
- 8 L/min. (Air)
- 8 L/min. (Air)
- Vacuum range:
- –300 - –600 mbar (adjustable)
- –300 - –600 mbar (adjustable)
- –300 - –600 mbar (adjustable)
- Weight:
- 3.4 kg (6.83 lbs.)
- 3.4 kg (6.83 lbs.)
- 3.4 kg (6.83 lbs.)