Tubing, Tubing Cartridges, and Accessories for Peristaltic Pumps, Welch®, Gardner Denver
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
For use with Welch® peristaltic pumps (see 84009-696 series).
Silicone tubing is flexible and translucent with good chemical and environmental resistance and very good electrical resistance. High-performance, flexible PVC tubing features great versatility and moderate chemical resistance and is ideal for use with peristaltic pump heads. Lok-n-Flow tubing cartridges allow easy replacement of worn tubing at the pump head without wasted materials.
Ordering information: The starter tubing kit 84009-704 includes 7.6m (25') of 4.8mm I.D. general PVC tubing; four hose clamps; and two each of 1.6mm (1/16"), 3.2mm (1/8"), and 4.8mm (3/16") I.D. silicone tubing cartridges.
Customers Who Bought This Also Bought