A propos de cet article
Vacuum ovens are designed for drying, curing, outgassing, aging, process control, and other applications which require elevated temperatures in reduced atmospheres, or vacuum/purge with nonflammable and inert atmospheres
Fiberglass insulation minimizes heat loss in walls and door. Stainless steel interiors provide resistance against corrosive materials. Oven exteriors are available in stainless steel or white painted finish. Ovens with stainless steel exteriors feature solid outer door with inner window door.
Caution: Not suitable for use with hazardous vapors.
All ovens feature viewing window, high temperature silicone gaskets, built-in over-temperature protection, and a fully flexible vacuum/purge/release system. Front-mounted single setpoint digital microprocessor control with LED readout simultaneously displays actual temperature and setpoint.
Spécifications
- Catalog No:
- 52348-360
- 52348-350
- Temperature range:
- Amb. +6…+260 °C (500 °F)
- Amb. +6…+260 °C (500 °F)
- Temperature uniformity:
- ±3.5% of setpoint, nominal
- ±3.5% of setpoint, nominal