Drum Cradle, Wesco
Fournisseur: Wesco
Certificats
A propos de cet article
Economical, convenient way to tilt, drain and store 208.2 L (55 gal.) drums
Easy for one person to use. Drum distance to floor: 38 cm (15").
Outboard style has four rigid polyolefin wheels. Inboard style features two swivel, two rigid polyolefin wheels.
Packaging: Shipped unassembled for quick, easy assembly.
Customers Who Bought This Also Bought