Refractometer, Industrial Fluid Tester, Reichert
# de catalogue 55680-389
Fournisseur: Reichert Technologies
Spécifications
- Resolution:0.5
- Refractive index:1.3330–1.3860
- Cat. no.:55680-389
- Supplier No.:13754000
- Scale Range:0 - 30
Spécifications
A propos de cet article
Automatic temperature compensation for immediate, accurate analysis without referencing temperature conversion charts
Easy-to-read scale instantly measures cutting fluids, water soluble coolants, oil/water mixtures, cleaners, plating baths, and many other types of industrial solutions. Includes instructions for maintaining optimum fluid/water ratios. Durable, water-resistant design. Scale is measurement of an arbitrary scale based on coolants for metal working.
Customers Who Bought This Also Bought