Orbitron Rotators, Boekel Scientific
Fournisseur: Boekel
Certificats
A propos de cet article
Gentle, uniform, low-foam mixing action suitable for immunoassays, hybridizations, mixing blood samples, coupling affinity chromatography media, and DNA extractions
Rotator I features a fixed speed of 20rpm and tilt of 23°. Rotator II features variable speed of 6–23rpm and tilt of 0–23°.
Compact unit has 30.5cm (12") square platform. Dimpled pad on platform accepts a variety of sizes of tubes simultaneously with no need for racks or clamps. Pad is removable for easy cleaning and test tube transport.
Ordering information: Rotator I is supplied with one dimpled pad. Rotator II is supplied with one dimpled pad and one flat, nonskid pad. Accessory tray kit includes a 20.3cm (8") platform, one dimpled pad, and one flat, nonskid pad.
Spécifications
- Catalog No:
- 35821-000
- 56261-508
- Dimensions:
- 19W x 17.8D x 12.7H cm (71/2 x 7 x 5")
- 19W x 17.8D x 12.7H cm (71/2 x 7 x 5")
- Weight:
- 3.4 kg (7.5 lbs.)
- 3.4 kg (7.5 lbs.)
- Electrical:
- 115V, 0.4A
- 115V, 0.4A