Spécifications
- Description:Hybridization Oven
- Electrical:115V, 230W, 50/60Hz
- Temperature range:5° above ambient to 70°C (158°F)
- Overall dimensions:41H x 32W x 37D cm (16 x 121/2 x 141/2")
- Chamber dimensions:27H x 29D x 29W cm (101/2 x 111/2 x 111/2")
- Shaking speed:8 to 25 strokes per minute
- Temperature stability:±0.2° at 65°C
- Weight:11 kg (24 lbs.)
- Cat. no.:35821-077
- Supplier No.:136400
Spécifications
A propos de cet article
This hybridization incubator is ideal for hybridizing and prehybridizing membranes used in Southern and Northern blotting techniques
Three 22x20x1.3cm (821/32x77/8x1/2") shelves, spaced 6.35cm (21/2") apart, accommodate dishes or heat-sealed bags. Shelves are removable to increase space for taller vessels. Equipped with a gasket-sealed, metal and acrylic see-through door.
The control panel features temperature selection buttons; an easy-to-read LCD temperature display; an independent, calibrated, safety thermostat; and a speed adjustment knob.
The gentle rocking motion provides washes that are less messy than a shaking water bath and more convenient than a shaker inside a conventional incubator. The small footprint conserves valuable bench space.