VWR® Screw-Thread Vials 8-425
Supplier: VWR International
Certificates
About this item
Standard opening screw-thread vials are manufactured from borosilicate glass and have a maximum fill volume of 1.8 ml.
- Borosilicate glass
- Screw caps with preattached septa
Closures are black and have preattached septa.
Customers Who Bought This Also Bought