KIMAX® Burets, Serialized and Certified, Class A, Three-Way PTFE Stopcock, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Each KIMAX® precision bore buret has a permanent unique serial number and durable white enameled scale
Manufactured according to ASTM E287 Class A requirements.
Fill through the stopcock using 0.25" I.D. flexible tubing. Stopcock [PS] 2.
Ordering information: Shipped complete with KIM-KAP® dust cap and Certificate of Graduation Accuracy. Shipped in NSTA approved packaging.
Customers Who Bought This Also Bought