Standard Ball Valve Lockout, Brady®
# de catalogue CA89011-764
Fournisseur: Brady Worldwide
Spécifications
- For Use With:Ball Valve Sizes from 6.35 to 25.4mm (0.25 to 1")
- Color:Red
- Cat. no.:CA89011-764
- Supplier No.:65666
Spécifications
A propos de cet article
Ideal for securing valve handles in the on or off position
- Easy to use, single-piece design
- For use on ball valve sizes from 6-25mm (1/4-1")
- Accepts 7mm (9/32") padlock shackles
Easy to use, single-piece design makes the standard ball valve lockouts the perfect tool for securing valve handles in the off position.
The hole diameter accepts 7.14mm (⁹⁄₃₂") padlock shackles.
Customers Who Bought This Also Bought