Freezing Point Apparatus, Koehler
# de catalogue 52932-022
Fournisseur: Koehler
Small Business Enterprise
Spécifications
- Cat. no.:52932-022
- Shipping Weight:7.3 kg (16 lbs.)
- Dimensions:28Lx23Wx82.5H cm (11x9x32.5")
- Supplier No.:K29700
Spécifications
A propos de cet article
Cools aviation gasoline and aircraft turbine fuel samples to determine the temperature at which hydrocarbon crystals appear
Conforms to ASTM D2386 specifications.
Consists of unsilvered vacuum flask, jacketed sample tube, brass packing gland, stirrer, stopper, clamps and stand. Uses acetone or alcohol cooled by solid carbon dioxide as a cooling medium.
Ordering information: Complete with line cord and switch. Mechanical stirrer motor not included.
Customers Who Bought This Also Bought