PYREX® Melting Point Tube, Corning
# de catalogue 89091-274
Fournisseur: Corning
Spécifications
- Length:150 mm
- O.D.:25 mm
- Description:Thiele Melting point tube
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89091-274
- Supplier No.:9540-25
Spécifications
A propos de cet article
For determining the melting point of organic compounds.
Customers Who Bought This Also Bought