Universal Grinding Mills, IKA® Works
Fournisseur: IKA Works
Certificats
A propos de cet article
Suitable for grinding up to 250mL of hard, brittle substances
Maximum grain size is 6 to 7mm. Unit features an exchangeable grinding chamber with handles. Exchangeable cutters for universal dry crushing. Easy to clean. Cast aluminum housing. Output: 225 watts.
Ordering information: Supplied with stainless steel blade, grinding chamber and lid, cord, and plug.
Spécifications
- Catalog No:
- 33998-048
- 33998-050
- Dimensions:
- 17W x 17L x 35H cm (63/4 x 63/4 x 133/4")
- 17W x 17L x 35H cm (63/4 x 63/4 x 133/4")
- Weight:
- 6.5 kg (141/2 lbs.)
- 6.5 kg (141/2 lbs.)