ContactUsLinkComponent
VWR® Premium Disposable Microtome Blades
VWR® Premium Disposable Microtome Blades
# de catalogue 95057-832
Fournisseur:  VWR International
VWR Low Profile Coated Microtome Blades
VWR® Premium Disposable Microtome Blades
# de catalogue 95057-832
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Triple-Facet, Coated
  • Cat. no.:
    95057-832
Spécifications
A propos de cet article
Designed to fit standard blade holders, premium disposable blades are ideal for all routine and specialty histology applications.
Designed to fit standard blade holders, premium disposable blades are ideal for all routine and specialty histology applications. Low profile triple-facet blades feature an extra-sharp cutting edge for thinner sections that will cut down to 2µm, and a fluoropolymer resin coating to ensure a consistent cutting edge and increase blade life.
  • Heavy gauge, high-quality carbon steel construction
  • High-profile blades
  • Ultra-thin sectioning
  • Sections jump off edge forming beautiful ribbons
Low-profile triple-facet blades feature an extra-sharp cutting edge for thinner sections that will cut down to 2µm, and a fluoropolymer resin coating to ensure a consistent cutting edge and increase blade life.
Low-profile triple-facet blades feature an extra-sharp cutting edge for thinner sections that will cut down to 2µm, and a fluoropolymer resin coating to ensure a consistent cutting edge and increase blade life.
High-profile coated blades are ideal for all routine and specialty histology applications. Sections as thin as 2µm are possible with these blades, and they also feature a fluoropolymer resin coating to ensure a consistent cutting edge and increase blade life. Thick high-profile blades eliminate chattering when sectioning hard specimens. This blade will not bend during cutting, eliminating the thick/thin sectioning problem often found when cutting hard tissue. These blades are ideal for sectioning bone, eyes, and other hard tissue. They are also an outstanding choice for industrial purposes and frozen sections.
Ordering information: Blades are supplied in an ejector dispenser pack equipped with a storage compartment to safely discard spent blades.
Customers Who Bought This Also Bought