Single Compartment Sinks with Ledgeback, Just Manufacturing
Fournisseur: Just Manufacturing
Certificats
A propos de cet article
Fabricated of Type 304, 18-8 stainless steel with polished interior and top surfaces
Self-rimming sinks feature slightly raised edge perimeter to prevent water from running onto countertop. Center-punched bowl receives cup strainer. Faucet ledge width: 11.4 cm (41/2"). Sink 58200-552 has ledge width of 8.8 cm (31/2").
All sinks thoroughly sound deadened.
Customers Who Bought This Also Bought