Double Compartment Sinks with Ledgeback, Just Manufacturing
Fournisseur: Just Manufacturing
Certificats
A propos de cet article
Fabricated of Type 304, 18-8 stainless steel with polished interior and top surfaces.
- Self-rimming
- Punched center receives cup strainer
All sinks are thoroughly sound deadened. Faucet ledge width: 11.4 cm (4½").
Customers Who Bought This Also Bought