Burundum™ Cylinders, U.S. Stoneware
Fournisseur: US Stoneware
Certificats
A propos de cet article
For wet or dry processes, the cylindrical shape provides additional contacts for faster, finer grinding
The non-porous surface is hard and chip-resistant, non-contaminating. High specific gravity (3.4) ups “packing density” for most efficient mill use. The smaller size is for liquid dispersion, and the larger size for crushing. Hardness is 9 on the Mohs scale.
Customers Who Bought This Also Bought