Stainless Steel Slanted Storage Rack, Bandy
# de catalogue 24450-010
Fournisseur: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Load Capacity:800 lbs.
- Supplier No.:WRK60003
Spécifications
A propos de cet article
Storage rack features rod-top shelves sloped at an 8° angle for safe storage of work-in-process
Constructed of 16-gauge, type 304 stainless steel, completely electropolished. Adjustable leveling feet.
Customers Who Bought This Also Bought