ContactUsLinkComponent
Ergonomic Drum Handler, Model DM-1100, Wesco
Ergonomic Drum Handler, Model DM-1100, Wesco
# de catalogue 21002-012
Fournisseur:  Wesco
Ergonomic Drum Handler
Ergonomic Drum Handler, Model DM-1100, Wesco
# de catalogue 21002-012
Fournisseur:  Wesco
Spécifications
  • Dimensions:
    121.9H x 87.6L x 91.4W cm (48 x 341/2 x 36") legs retracted
    121.9H x 94L x 104.1W cm (48 x 37 x 41") legs extended
  • Cat. no.:
    21002-012
  • Shipping Weight:
    181 kg (400 lbs.)
  • Load Capacity:
    295 kg (650 lbs.) legs retracted
    499 kg (1100 lbs.) legs extended
  • Supplier No.:
    240150
Spécifications
A propos de cet article
Drum handler with hydraulic jack is ideal for lifting, transporting, and placing steel, poly, or fiber drums onto spill pallets or standard pallets.
  • Eliminates need to touch drum while raising, transporting, or lowering it
  • Unique spring-loaded clamp holds any drum rim 4.8mm (3/16") or higher, even over rough terrain
  • Accommodates most standard 114L (30gal.), 208L (55gal.), and 322L (85gal.) drums, and enables 208.2L (55gal.) drums to be easily raised 50.8cm (20") above the floor
  • Also features floor lock and swiveling phenolic casters for easy movement and steering
  • Rise/stroke: 2.5cm (1")
A unique spring-loaded clamp holds any drum rim 4.8mm (3/16") or higher, even over rough terrain. No need for the operator to touch the drum to raise, transport, or lower it. Drum handler can accommodate most standard 114L (30gal.), 208L (55gal.), and 322L (85gal.) drums, and enables 208.2L (55gal.) drums to be easily raised 50.8cm (20") above the floor. Drum handler also features swiveling phenolic casters for easy movement and steering, and a floor lock.
Customers Who Bought This Also Bought