Drum Storage Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Drum storage cabinets enable easy storage of flammable liquids in 208.2 L (55 gal.) drums.
- Two-door cabinets are available in horizontal or vertical models
- All cabinets have a large “FLAMMABLE – KEEP FIRE AWAY” label
The horizontal model featuring tracks for easy placement or removal of drum cradle. The vertical models have ball-bearing roller bottoms. Cabinets are constructed of double-wall, 18-gauge galvanized steel and include a flush-mounted locking handle, three-point self-latching closure, and full-length heavy-gauge piano hinges. Cabinets 15001-684 and 14226-900 are supplied with adjustable 18-gauge galvanized steel shelves. Self-close cabinets feature a fusible link that melts at 71 °C (165 °F) allowing the doors to automatically close and latch.
Certifications: Meet NFPA Code 30 specifications. All cabinets FM approved.
Spécifications
- Catalog No:
- 15001-682
- 14226-898
- 15001-680
- 14226-900
- 15001-684
- 14226-896
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified