Protector® Vacuum Pump Storage Cabinets, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Storage cabinets provide enclosed space for vacuum pump storage and can also support Labconco® laboratory hoods.
- Cabinets include a manual-closing, non-locking, single door available with right or left hinge opening
Storage cabinets provide enclosed space for vacuum pump storage and can also support Labconco® laboratory hoods.
Cabinets are constructed of white epoxy-coated steel and include one 20.3 cm (8") filler panel to increase cabinet’s overall depth from 55.9 cm (22") to 76.2 cm (30"), for installation flush to wall.
Ordering information: Cabinets are supplied with four leveling feet; one 20.3cm (8") filler panel; rear cutouts for vacuum pump tubing; and one 115V, 60Hz electrical duplex receptacle. The receptacle requires wiring to provide power to the vacuum pump. A work surface (sold separately) is required for these cabinets.
Customers Who Bought This Also Bought