Cylinder Lockers, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
Safely secure gas cylinders, including liquefied petroleum gas (LPG) and oxygen cylinders, from theft and unauthorized access
- Sloped roof to shed water and ice
- Solid top to reinforce stability
- Warning and full/empty cylinder labels
- Extruded framework for extra strength
- Padlock lug for security
- Pre-drilled holes at each corner for anchoring
Horizontal lockers feature easy-to-use racks that position cylinders securely and can hold 9.1 kg (20 lbs.) or 15 kg (33 lbs.) cylinders. Vertical lockers feature a heavy-duty floor plate to keep cylinders clear of ground and adjustable plated steel chains to hold cylinders in place.
Welded aluminum lockers will not rust, corrode, or discolor.
Certifications: Lockers comply with NFPA 58 and OSHA 1910.110 standards.
Customers Who Bought This Also Bought