VWR® Atlas Overhead Storage Cabinets with Hinged Doors
Fournisseur: VWR International
Certificats
A propos de cet article
Steel cabinets are used in place of open shelving when enclosed storage and/or resistance to dust is required.
- Hinged doors
- Color: Light neutral
For double-sided storage without pass-through, two cabinets of the same size may be mounted back to back.
For use with VWR® Atlas mobile carts/mobile laboratory benches.
Ordering information: Supplied with two adjustable shelves and mounting hardware. Hardware required for back-to-back installation not included.
Customers Who Bought This Also Bought