Automatic Aniline Point Apparatus, Koehler
Fournisseur: Koehler
Certificats
A propos de cet article
Used to characterize pure hydrocarbons and to indicate the aromatic content of hydrocarbon mixtures
Apparatus performs hands-free aniline point and mixed aniline point determinations automatically by means of a modified thin film technique (ASTM D611 Method E). The sample is heated by a platinum immersion heater and the aniline point is detected photoelectrically. Temperature is displayed on a large LED indicator. Built-in pressure regulator and solenoid valve permit quicker cooling cycles and determination of subambient aniline point temperatures. Cabinet exterior surfaces have a chemical-resistant polyurethane enamel finish. Standard borosilicate glass test cell with drain is provided.
Certifications: Conform to specifications: ASTM D611, IP 2, ISO 2977, DIN 51775, FTM 791-3601, NF M 07-021.
Spécifications
- Catalog No:
- 37000-018
- 37000-014
- Temperature range:
- 0 to 999.9 °C (Display)
- 0 to 999.9 °C (Display)
- Testing range:
- 0 to 150 °C (32 to 302 °F)
- 0 to 150 °C (32 to 302 °F)
- L×W×H:
- 37 x 22 x 53 cm (141/2 x 81/2 x 203/4")
- 37 x 22 x 53 cm (141/2 x 81/2 x 203/4")
- Shipping weight:
- 21 kg (46 lbs.)
- 21 kg (46 lbs.)