A propos de cet article
Regulators have a brass barstock body and are suitable for a variety of applications where pressure rise is not critical, such as high-purity gas applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems.
- Brass body
- Stainless steel diaphragm
- Sintered filter
- PTFE seat design
A stainless steel diaphragm eliminates contamination from diffusion or outgassing
Note: For noncorrosive gases only.
A one-piece PTFE encapsulated seat design includes a sintered filter to protect the seat from particulate contamination. The 1x10-9cc/sec. inboard helium leak rate maintains gas purity levels. Front or back panel mountable.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 55850-604
- 55850-606
- 55850-608
- 55850-610
- 55850-612
- 55850-614
- 55850-616
- 55850-618
- 55850-620
- 55850-600
- 55850-602
- Max. inlet pressure:
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- 3000 psig
- Body outlet:
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- Diaphragm:
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Type 316L Stainless Steel, 4.3 cm (111/16")
- Dual scale gauges:
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- 5 cm (2") psi/bar
- Outlet type:
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting
- Diaphragm Valve with 0.6 cm (1/4") Tube Fitting