A propos de cet article
Regulators have a brass body and are suitable for a wide variety of applications where small changes in the gas delivery pressure will not degrade performance.
- Brass body
- Easy-to-read gauges
- Large diaphragm ensures constant delivery pressure
- Neoprene seat
- Sintered metal inlet filter eliminates particulates
Gauges are easy to read
Note: For noncorrosive gases only. Use high-pressure hoses on regulators 55850-275, 55850-277, and 55850-270.
Large diaphragm ensures constant delivery pressure. Regulators with neoprene diaphragms have a valve, which closes with pressure for safety. Regulators with stainless steel diaphragms are not supplied with a valve. Particulates are eliminated from gas stream with sintered metal inlet filter.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 55850-200
- 55850-255
- 55850-270
- 55850-275
- 55850-245
- 55850-250
- 55850-205
- 55850-277
- 55850-215
- 55850-220
- 55850-225
- 55850-230
- 55850-235
- 55850-240
- 55850-260
- Body outlet:
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- Diaphragm:
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Neoprene, 7 cm (23/4")
- Gauge:
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- 5.1 cm (2")
- Valve outlet:
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT
- 0.6 cm (1/4") NPT