A propos de cet article
The sample is evaporated and condensed under controlled conditions, with measurements of the temperatures at which various percentages are recovered and/or the percentages recovered at specified temperatures
Distillation apparatus are suitable for groups 0, 1, 2, and 3: motor and aviation gasolines, aviation turbine fuels, naphthas, kerosenes, distillate fuels, natural gasoline, liquid hydrocarbon mixtures, and other petroleum products. The 1000W heater unit and condenser are stainless steel.
Distillation apparatus are suitable for groups 0, 1, 2, and 3: motor and aviation gasolines, aviation turbine fuels, naphthas, kerosenes, distillate fuels, natural gasoline, liquid hydrocarbon mixtures, and other petroleum products. The 1000W heater unit and condenser are stainless steel.
Group 4 apparatus are additionally suitable for testing Grade No. 2 fuel oil, Grade No. 2-D diesel fuel oil, gas oils, and other distillates requiring condenser bath temperatures up to 60°C (140°F). Group 4 units feature a 300W copper immersion condenser heater.
Units feature fully insulated condenser and heater units with stepless electronic control, flask support platform, viewing window, and rack and pinion heater-elevation mechanism with push-turn control knob.
Spécifications
- Catalog No:
- 37000-284
- 37000-282
- 37000-270
- 37000-278
- 37000-276
- 37000-272
- 37000-280
- L×W×H:
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- 39 x 46 x 50 cm (151/4 x 181/4 x 191/2")
- Shipping weight:
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)
- 29.5 kg (65 lbs.)