Ramsbottom Carbon Residue Apparatus, Koehler
Fournisseur: Koehler
Certificats
A propos de cet article
Provides an indication of the deposit-forming tendencies of fuels by determining the volume of carbon residue left after evaporation and pyrolysis of a sample oil
Cast-iron block type coking furnace features microprocessor temperature control with °C/°F switchable digital set point and display. Stainless steel cabinet is heavily insulated with a three-layer refractory top for excellent heat retention. Built-in control circuit automatically interrupts power if the block temperature exceeds the programmed cutoff point.
Certifications: Conforms to specifications: ASTM D524, D6074; IP 14; ISO 4262; FTM 791-5002.
Spécifications
- Catalog No:
- 37000-184
- 37000-182
- Sensitivity:
- ±1°C (±2°F)
- ±1°C (±2°F)
- Max. temperature:
- 650°C (1200°F)
- 650°C (1200°F)
- L×W×H:
- 41 x 55 x 37 cm (16 x 211/2 x 141/2")
- 41 x 55 x 37 cm (16 x 211/2 x 141/2")
- Shipping weight:
- 33.1 kg (73 lbs.)
- 33.1 kg (73 lbs.)