KIMAX® Cold Test Jar, Cloud and Pour Points, Kimble Chase
Catalogue # 89002-138
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:35 mm (13/8"): 35 mm (13/8")
- Height:125 mm (4⅞")
- Cat. no.:89002-138
- Supplier No.:32501-99
Spécifications
À propos de cet article
Used to determine the cloud point of petroleum oils and the point at which chilled, undisturbed oils will pour
Flat bottom jar features a reinforced bead at open end, a marking spot, and a graduation ring at 54mm (21/8") from inside bottom.
Certifications: Meets ASTM D2500 and D97 requirements.