VWR® Atlas Single Uprights
Supplier: VWR International
Certificates
About this item
Counter-mounted Atlas upright supports are designed to bolt directly to a new or existing countertop to provide infinitely adjustable shelving above the bench.
- Light neutral supports with black metal inserts
These Atlas single uprights are built of durable materials.
Ordering information: Supplied with mounting hardware. Uprights 152.4 cm (60") and higher are supplied with a ceiling trim ring.
Customers Who Bought This Also Bought