VWR® Atlas Slotted Uprights
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Slotted uprights are bolted to a standard Atlas upright to modify the upright to adjustability on 2.5 cm (1") increments.
- Color: Light neutral
Slotted uprights are to be installed on the inside of each column upright.
Ordering information: Each slotted upright is provided with mounting hardware and plugs to cover mounting holes.
Customers Who Bought This Also Bought