A propos de cet article
Ideal for storage of flammable liquids and solvents near laboratory benches, fume hoods, and other work areas.
- Available with manual or self-closing doors
- The self-close doors are equipped with a fusible link that automatically closes the door if the link melts due to fire
- The sides, top, and bottom are double-walled with 3.8 cm 1½" air space between walls
Additional features include grounding attachments, three-point key lock on doors, 5 cm (2") raised leakproof doorsills, and four leveling feet. The adjustable shelves are supported by four brackets and can hold up to 226.8 kg (500 lbs.). Undercounter cabinets 56610-270 and 56610-272 have a recessed kickplate for ergonomic working conditions.
Maximize the storage capacity of schools, pharmacies, production areas, and laboratories. Cabinets are constructed of 18-gauge steel and finished in a caution yellow powder-coat paint with 'Flammable – Keep Fire Away' in red lettering on the front. Label is in three languages (English, Spanish, and French.)