Double-Bowl Sinks, Polyolefin, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
Double-bowl sinks are constructed of a unique molded thermoplastic VSP polyolefin.
- Stainless steel rims available for flush mount
- One-piece construction provides maximum contamination protection
- Steel rims available for under counter mount
Customers Who Bought This Also Bought