Corner Drain Sinks, Polyolefin, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
One-piece, single-bowl sinks with corner drain.
- Polyolefin unit allows easy cleaning and reduced glassware breakage
Color: black.
Customers Who Bought This Also Bought