Deck-Mounted Pure Water Gooseneck Faucets, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted pure water faucets feature tin-lined, chrome-plated brass or all-PVC construction.
Tin-lined faucets are provided with self-closing control and 6 mm (1/4") NPT tin-lined tail piece on inlet. All-PVC faucet is provided with 1.3 cm (1/2") IPS PVC shank, locknut, and washer on inlet.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought