Deck-Mounted Pure Water Faucets, Epoxy-Coated, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted pure water faucets are made of white epoxy/powder-coated brass.
Tin-lined faucet is provided with self-closing control and 6mm (1/4") NPT tin-lined tail piece on inlet. Polypropylene-lined faucets are provided with 1 cm (3/8") compression tube adapter on inlet.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought