VWR® Contour™ Knee Space Filler Panels
Fournisseur: VWR International
Certificats
A propos de cet article
Designed to close off the space between the floor and the bottom of sitting- or standing-height aprons
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
- Light neutral color
Panels are also used to conceal piping in furniture assemblies that include aprons. Two filler angles are included for mounting.
Customers Who Bought This Also Bought